Learn More
JDM SCIENTIFIC LLC Smart Rack starter set for Life Science
Supplier:  JDM SCIENTIFIC LLC 1510
Encompass_Preferred
The SmartRack for Lifescience has been specially designed for work in the fields of biology, biomedicine, biotechnology as well as special fields of biology and for handling liquids in small quantities and preparing and storing samples for later analysis. It includes the following components (item number of each component in brackets for better understanding): 16 components:- 1 x SmartRack (Item no. 01704)- 1 x Holder for bin liner (Item no. 01144)- 1 x Storage box (Item no. 01142)- 1 x Storage box insert - for small accessories (Item No. 01148)- 1 x Storage box insert - for microreaction vessels, 20 x 2 ml (Item no. 01130)- 1 x Storage box insert - for centrifuge tubes, 6 x 15 ml, 4 x 50 ml (Item no. 01102)- 2 x Holder for laboratory screw top bottles, 100 - 1000 ml (GL45) (Item no. 01122)- 2 x Holder for well plates (Item no. 01125)- 1 x Holder for wash bottles, for 500 ml (up to Ø 75 mm) (Item no. 01146)
Catalog No. 50-255-8511
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.