Learn More
LaMotte SMART Spectro Spectrophometer and SMART 3 Colorimeter Reagent Systems
For water analysis in the field or in the lab
Manufacturer: LaMotte 3642SC
Description
- High-quality reagents manufactured from ACS raw materials for high reproducibility
- All systems are designed to overcome test interferences for fast and dependable results
Specifications
Ammonia Nitrogen | |
SMART Spectro Spectrophometer and SMART 3 Colorimeter | |
50 |
0 to 4ppm | |
Water testing and environmental studies | |
Nesslerization |