Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ SMART™ Spectro System COD Reagents
SDPSCI_ED

Catalog No. S72413 Shop All LaMotte Company Products
Change view
Click to view available options
Range:
0 to 15,000ppm
0 to 1500ppm
0 to 150ppm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S72413 0 to 1500ppm
S72414 0 to 15,000ppm
S72412 0 to 150ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S72413 Supplier LaMotte™ Supplier No. 0076SC
Add to Cart
Add to Cart

A precise and easy way to measure critical COD levels.

Measure low, medium or high levels of Chemical Oxygen Demand (COD) using a mercury reagent system approved by the Environmental Protection Agency (EPA).

  • Comes in 25 vials to a case
  • Requires COD Heater Block, not included

Specifications

Certifications/Compliance EPA
For Use With (Equipment) LaMotte™ colorimeters
Range 0 to 1500ppm
Test Method Acid digestion
Length (English) 0.62 in.
Length (Metric) 1.6 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.