Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Smart-Vue™ Event Notification Hardware

Catalog No. SV509500LSB Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Smart-Vue Audible and Visual Alarm, 915MHz
Smart-Vue Voice and Fax Alert
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
SV509500LSB Smart-Vue Voice and Fax Alert
SV500500LSB Smart-Vue Audible and Visual Alarm, 915MHz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SV509500LSB Supplier Thermo Scientific™ Supplier No. SV509500LSB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Receive immediate notification by means of your preferred alert method, including audible/visual siren, fax, telephone, email or text messaging.

Receive immediate notification by means of your preferred alert method, including audible/visual siren, fax, telephone, email or text messaging. Thermo Scientific™ Smart-Vue™ Event Notification Hardware delivers the continuous monitoring, secure data logging and automatic audit trails required to comply with SOP, cGxP and 21 CFR part 11 regulatory standards.

Local RF Audible and Visual Alarm

  • Universal (100V.240V) to 12 volt power supply
  • 3.6V battery
  • Velcro
  • Hardware
  • Customer reference letter

Voice and Fax Alert

  • USB 1.0 or 2.0 powered
  • Fits standard RJ11 telephone line

Ordering Information:

Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.

TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Fax
Description Smart-Vue Voice and Fax Alert
Includes USB 1.0 or 2.0-powered voice/fax alert
Certifications/Compliance Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application) Cold Storage
For Use With (Equipment) Standard RJ11 Telephone Line
Operating Frequency 915 MHz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.