Learn More
Thermo Scientific™ Smart-Vue Pro Accessories
Catalog No. 11676460
Change view
Click to view available options
Type:
Adapter
Battery
For Use With (Equipment):
2 or 3 Wire Generic PT100 Probes
Smart-Vue Classic PT100 Probes
Smart-Vue Pro Duo or Quatro Module
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
11-676-460 Adapter 2 or 3 Wire Generic PT100 Probes
SVACCBAT002 Battery Smart-Vue Pro Duo or Quatro Module
11-676-461 Adapter Smart-Vue Classic PT100 Probes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-676-460 Supplier Thermo Scientific™ Supplier No. SVPADPPT1001
Add to Cart
Add to Cart
This item is not returnable. View return policy
Customize your Smart-Vue Pro remote monitoring system with accessories to meet your specific needs.
Choose from an assorment of Smart-Vue Pro accessories including power supplies, cables and mounting hardware, antenna and audible alarms to customize your remote monitoring system as well as ensure system integrity.
Compatible with Smart-Vue Pro remote monitoring system.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Calibration Calibration parameter are stored in the adapter. If calibrated sensors are connected a recalibration of the whole chain is recommended.
Description Smart-Vue Pro adaptor for use with 2 or 3 wire generic PT100 probes. The adapter connects directly to a Smart-Vue Pro Duo or Quatro module
For Use With (Equipment) 2 or 3 Wire Generic PT100 Probes
Measurement Range Depends on the sensor connected at the adapter
Length (English) Cable 3.6 ft.
Weight (Metric) 150 g (Including shipping box)