Learn More
Eppendorf™ SmartBlock DWP500
Catalog No. 05414305 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Deepwell plates, 96/500 μL
Catalog No. For Use With (Equipment)
05-414-305 Deepwell plates, 96/500 μL
1 options
This item is not returnable. View return policy
Catalog No. 05-414-305 Supplier Eppendorf™ Supplier No. 5316000004
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermoblock for DeepWell Plate supports optimal temperature transfer within your sample.
  • For Eppendorf Deepwell Plate 96/500 μL
  • Optimized block design supports optimal temperature transfer within your sample to receive excellent results
  • Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
  • Every SmartBlock is individually inspected in production for temperature accuracy and is delivered includes serial-number-specific certificate for documentation purpose
  • Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept)
  • Automatic block recognition for safe handling
  • Eppendorf QuickRelease™ technology enables fast and simple block exchange - no need for tools
  • Limited to ThermoMixer C and ThermoStat C
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Deepwell plates, 96/500 μL
Type Block
Includes Lid