Learn More
JDM SCIENTIFIC LLC SmartCase
Supplier:  JDM SCIENTIFIC LLC 02101
Encompass
The SmartCase is designed to transport and store the SmartRack, a modular a modular organization system for laboratory workstations. Its aluminium case safely protects a SmartRack inside and its contents from light and other environmental influences. The SmartCase consists of a lower part with four castors into which a SmartRack can be placed. The upper part of the SmartCase has a cushion on the inside, which serves to fix the SmartRack in the SmartCase as soon as the SmartCase is closed by the tension locks. This prevents the SmartRack from slipping or tipping over. The SmartCase has a self-acting valve system which can be used to change the atmosphere inside the SmartCase by means of an external gas supply. This allows, for example, the temporary generation of a low-oxygen atmosphere in the SmartCase, which creates an additional safety level when transporting oxygen-sensitive, highly flammable substances or substances that spontaneously ignite in air.
Catalog No. 50-255-8517
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.