Learn More
Wanzl SmartCell™ Angled Basket
Description
- Baskets are available in a flat configuration or can be angled for easier access.
- Adjustable dividers can be added to customize
Specifications
Specifications
| Color | Chrome |
| Material | Steel |
| Dimensions (L x W) | 18 x 60 in. |
| For Use With (Application) | Shelving |
| Shelf Type | Angled Basket |
| Width (English) | 60 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.