Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wanzl SmartCell™ Post Mount Divider

Catalog No. 11222064
Change view
Click to view available options
Width (English):
10 in.
18 in.
24 in.
46 in.
48 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
11-222-064 46 in.
11-222-066 10 in.
11-222-062 18 in.
11-222-063 24 in.
11-222-065 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 11-222-064 Supplier Wanzl Supplier No. T.HPM46DV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Post mount dividers allow for easy custom configuration of mobile carts for greater mobility and visibility.

  • Allow for custom compartmentalization of the SmartCell™ basket system.
  • Large and small articles alike are stored in separate compartments, allowing for easy identification and quick retrieval.

Specifications

Color Chrome
Material Steel
For Use With (Application) Shelving
Shelf Type Divider
Width (English) 46 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.