Learn More
Wanzl SmartCell™ Shelving 3-Position Label Holder
Description
- Accessory for SmartCell™ Shelving
- 3 in. 3-Position Label Holders
- 50/Bag
Specifications
Specifications
| Color | Red |
| Length (English) | 3 in. |
| Includes | 50 Label Holders |
| For Use With (Application) | Shelving |
| With Casters | No |
| Material | Plastic |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.