Learn More
Wanzl SmartCell™ Shelving Dust Cover
Description
- Dust Cover
- Replacement part for SmartCell Shelving
Specifications
Specifications
| For Use With (Application) | Shelving |
| Dimensions (L x W) | 18 x 36 in. |
| Width (English) | 36 in. |
| Color | White |
| Includes | 1 Dust Cover |
| With Casters | No |
| Material | Vinyl |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.