Learn More
Clontech Labs 3P SMARTER PCR CDNA SYNTHESIS
SDP
Supplier:  Clontech Labs 3P 634925
Encompass
Synthesize full-length cDNA from ng of total RNA. Easy and efficient process maintains true gene representation. cDNA is immediately ready for downstream applications such as PCR, cloning, library construction & subtractive hybridization.
Catalog No. NC0230484
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.