Learn More
JDM SCIENTIFIC LLC SmartRack Mini
Supplier:  JDM SCIENTIFIC LLC 1707
Encompass_Preferred
The SmartRack was developed for use at manydifferent laboratory workstations and especially forwork in fume hoods. The SmartRack mini is speciallydesigned for work in safety cabinets.The SmartRack mini includes the following components:- a frame incl. a pair of feet- three slot rails each with handles- a drip tray
Catalog No. 50-255-8507
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.