Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580042 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580042 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580042 Supplier Invitrogen™ Supplier No. PA580042
  • $526.00
    $431.65 / Each of 1

    Save $94.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human HepG2 whole cell, human PANC-1 whole cell, human SK-OV-3 whole cell, human COLO-320 whole cell, rat testis tissue, mouse testis tissue. IHC: rat testis tissue, human testis tissue, mouse testis tissue. ICC/IF: A431 cell. Flow: A431 cell.

Core component of the SMC5-SMC6 complex, a complex involved in DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintenance via recombination in ALT (alternative lengthening of telomeres) cell lines and mediates sumoylation of shelterin complex (telosome) components which is proposed to lead to shelterin complex disassembly in ALT-associated PML bodies (APBs). Required for recruitment of telomeres to PML nuclear bodies. SMC5-SMC6 complex may prevent transcription of episomal DNA, such as circular viral DNA genome (PubMed:26983541). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMC6
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene SMC6
Gene Accession No. Q924W5, Q96SB8
Gene Alias 2810489L22Rik; 3830418C19Rik; AA990493; AU018782; AW742439; hSMC6; Kiaa4103; mKIAA4103; mSMC6; SMC protein 6; Smc6; SMC-6; SMC6 structural maintenance of chromosomes 6-like 1; SMC6L1; structural maintenance of chromosomes 6; structural maintenance of chromosomes protein 6; Structural maintenance of chromosomes protein 6-like protein
Gene Symbols SMC6
Host Species Rabbit
Immunogen E. coli-derived human SMC6L1 recombinant protein (Position: D205-E443).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313961, 67241, 79677
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.