Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMIFH2, Tocris Bioscience™

Catalog No. 440110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-011-0 10 mg
44-015-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-011-0 Supplier R&D Systems Supplier No. 4401/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

FH2 domain inhibitor; prevents actin nucleation

Inhibitor of formin homology 2 (FH2) domains. Prevents formin-mediated actin nucleation and barbed end elongation. Disrupts formin-dependent actin cytoskeletal structures in fission yeast and mammalian NIH 3T3 fibroblasts.

Chemical Identifiers

CAS 340316-62-3
Molecular Formula C15H9BrN2O3S
Molecular Weight (g/mol) 377.212
InChI Key MVFJHEQDISFYIS-XYOKQWHBSA-N
Synonym smifh2, 5e-1-3-bromophenyl-5-furan-2-ylmethylidene-2-sulfanylidene-1,3-diazinane-4,6-dione, smifh-2, 5e-1-3-bromophenyl-5-2-furylmethylene-2-thioxodihydropyrimidine-4,6 1h,5h-dione, 1-3-bromophenyl-5-2-furanylmethylene dihydro-2-thioxo-4,6 1h,5h-pyrimidinedione, 5e-1-3-bromophenyl-5-furan-2-ylmethylidene-2-thioxodihydropyrimidine-4,6 1h,5h-dione
PubChem CID 2258538
IUPAC Name (5E)-1-(3-bromophenyl)-5-(furan-2-ylmethylidene)-2-sulfanylidene-1,3-diazinane-4,6-dione
SMILES C1=CC(=CC(=C1)Br)N2C(=O)C(=CC3=CC=CO3)C(=O)NC2=S

Specifications

Quantity 10 mg
Formula Weight 377.21
Percent Purity >99%
Chemical Name or Material SMIFH2
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.