Learn More
Medchemexpress LLC HY-15694 5mg Medchemexpress, SMIP004 CAS:143360-00-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY156945MG
Encompass_Preferred
Medchemexpress, HY-15694 5mg SMIP004 CAS:143360-00-3 SMIP004 is a SKP2 E3 ligase inhibitor, which downregulates SKP2 and to stabilise p27. SMIP004 is a cancer cell selective apoptosis inducer of human prostate cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1769
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.