Description
- Meet ANSI Z87.1+ standards and provide 99.9% UVA/UVB/UVC protection
- Platinum frames have a self-adjusting nose pad with rubber fingers, which allows air flow and sweat to dissipate
- Polycarbonate lenses that help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Ergonomically dimensioned three-point extendable temples to adjust fit
- Cut-to-shape lens provides less distortion around the edges
Specifications
Specifications
| Lens Coating | Uncoated |
| Certifications/Compliance | Meets ANSI Z87.1+ standards for high impact (19826-00 meets ANSI Z87.1+2010 and 19826-01 meets ANSI Z87.1+2015) |
| Description | 3-Point fit |
| Lens Tint | Yellow |
| Product Type | Safety Glasses |
| For Use With (Application) | Eye protection |
| Frame Color | Black |
| Includes | Built-in hinge; Self-adjusting ergonomic rubber nose pad with rubber fingers |