Learn More
Ansell Smith & Wesson™ Magnum™ 3G Safety Glasses
Catalog No. 191300899 Shop All Ansell Products
Change view
Click to view available options
Frame Color:
Black
Includes:
Built-in hinge; Self-adjusting ergonomic rubber nose pad with rubber fingers
Lens Tint:
Clear
Yellow
Lens Coating:
Anti-fog
Uncoated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Includes Lens Tint Lens Coating
19-130-0899 Black Built-in hinge; Self-adjusting ergonomic rubber nose pad with rubber fingers Clear Anti-fog
19-130-0897 Black Built-in hinge; Self-adjusting ergonomic rubber nose pad with rubber fingers Yellow Uncoated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-0899 Supplier Ansell Supplier No. 19794
  • $499.51
    $313.65 / Dozen of 12
    Save $185.86
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Magnum 3G™ Safety Glasses are popular for use as shooting range glasses, gun glasses, gun range glasses or gun safety glasses
  • Meet ANSI Z87.1+ standards and provide 99.9% UVA/UVB/UVC protection
  • Platinum frames have a self-adjusting nose pad with rubber fingers, which allows air flow and sweat to dissipate
  • Polycarbonate lenses that help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
  • Ergonomically dimensioned three-point extendable temples to adjust fit
  • Cut-to-shape lens provides less distortion around the edges
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance Meets ANSI Z87.1+ standard
Description 3-Point fit
Lens Tint Clear
Product Type Safety Glasses
For Use With (Application) Eye protection
Frame Color Black
Includes Built-in hinge; Self-adjusting ergonomic rubber nose pad with rubber fingers