Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICU Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 1cc/23.5IU Kits

Catalog No. 22004201
Encompass
Change view
Click to view available options
Quantity:
100/Cs.
200/Cs.
Product Type:
Device, 2nd needle, Support kt
FP, NP Device, Support Kit
FP, NP Device, Support Kit pp
FP, PL Device, Support Kit, pp
Filter-Pro, Needle-Pro Device
Filter-Pro, Point-Lok Device
Needle Gauge:
23 to 25 G
25 G
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Needle Gauge
22-004-201 100/Cs. Device, 2nd needle, Support kt 23 to 25 G
22-004-204 200/Cs. Filter-Pro, Point-Lok Device 25 G
22-004-187 100/Cs. FP, PL Device, Support Kit, pp 25 G
22-004-186 100/Cs. FP, NP Device, Support Kit 25 G
22-004-185 200/Cs. Filter-Pro, Needle-Pro Device 25 G
22-004-184 100/Cs. FP, NP Device, Support Kit pp 25 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-004-201 Supplier ICU Medical Supplier No. 4640P1
Add to Cart
Add to Cart

Pro-Vent™ Plus 1 mL Arterial Blood Sampling syringes utilize lithium heparin with a concentration of 23.5 I.U. per mL. The specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference for more accurate and consistent results.The syringes are available with a broad choice of product features.

  • Advanced-formula dry lithium heparin
  • Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
  • Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
  • Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
  • Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
  • Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
  • Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage

Order Info

Sterile Tyvek packaging except as noted.

†Polybagged.

Specifications

Needle Gauge 23 to 25 G
For Use With (Application) Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results
Includes 1mL luer-slip syringe with Filter-Pro™ device, 25G x 0.62 in. preattached needle with Needle-Pro™ needle safety device, 23 G x 1 in. secondary needle with Needle-Pro™ needle safety device attached
Product Type Device, 2nd needle, Support kt
Quantity 100/Cs.
Packaging Sterile Tyvek™
Volume (Metric) 1 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.