Learn More
ICU Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 1cc/7IU Kits
Catalog No. 22024943
Encompass
Change view
Click to view available options
Quantity:
100/Cs.
200/Cs.
Product Type:
FP, PL Device, support kit, pp
Filter-Pro Device, Polybag
Filter-Pro, Needle Pro Device
Filter-Pro, Point-Lok Device
Packaging:
Polybag
Sterile Tyvek
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Packaging
22-024-943 200/Cs. Filter-Pro Device, Polybag Polybag
22-024-955 200/Cs. Filter-Pro, Point-Lok Device Sterile Tyvek
22-024-956 100/Cs. FP, PL Device, support kit, pp Sterile Tyvek
22-024-944 200/Cs. Filter-Pro, Needle Pro Device Sterile Tyvek
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-024-943 Supplier ICU Medical Supplier No. 46162
Add to Cart
Add to Cart
Pro-Vent™ Plus 1 mL Arterial Blood Sampling syringes utilize dry lithium heparin with a low concentration of 7 I.U. per mL. The low heparin concentration allows for healthcare professionals to obtain very small blood samples and still maintain the full range of blood gas, electrolyte, and metabolite testing with reduced heparin interference and extended clotting time.
  • Advanced-formula dry lithium heparin
  • Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
  • Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
  • Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
  • Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
  • Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
  • Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage
Order Info
Sterile Tyvek packaging except as noted.
†Polybagged.
Specifications
Needle Gauge 25 G
For Use With (Application) Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results
Includes 1mL luer-slip syringe with Filter-Pro™ device, 23.5 IU/mL concentration of dry lithium heparin as anticoagulant
Product Type Filter-Pro Device, Polybag
Quantity 200/Cs.
Packaging Polybag
Volume (Metric) 1 mL
Sterility Sterile