Learn More
ICU Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 3cc/70.5IU Kits
Description
- Advanced-formula dry lithium heparin
- Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
- Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
- Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
- Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
- Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
- Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage
Order Info
Sterile Tyvek packaging except as noted.
†Polybagged.
Specifications
Specifications
| Needle Gauge | 23 G |
| For Use With (Application) | Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results |
| Includes | 3mL luer-slip syringe with Filter-Pro™ device, 23G x 1 in. preattached needle with Needle-Pro™ needle safety device and iodophor prep pad, 23.5 IU/mL concentration of dry lithium heparin as anticoagulant |
| Product Type | FP, NP LS device, support kit |
| Quantity | 100/Cs. |
| Packaging | Sterile Tyvek |
| Volume (Metric) | 3 mL |
| Sterility | Sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.