Learn More
ICU Medical Pulsator™ Plus Dry Heparin Arterial Blood Sampling Kits
Shop All Smiths Medical PM Inc. ProductsDescription
Includes
3-cc or 5-cc Luer lock syringe and Filter-Pro™ device in polybag or Tyvek packaging.
Pulsator™ Plus 3 mL arterial blood sampling syringes utilize lithium heparin with a concentration of 23.5 I.U. per mL. The specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference with electrolyte values, for more accurate and consistent results. The syringes are available with a choice of product features.
- Pulsating effect allows for precise positioning in arterial lumen
- Syringe response to arterial pressure helps ensure arterial sample
- Rapid fill minimizes patient discomfort
- 3mL arterial blood-sampling syringes are easily aspirated during low blood pressure or code situation
- Needles with different gauges and lengths
- Utilize lithium heparin with a concentration of 23.5 IU/mL
- Specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference for more accurate and consistent results
Order Info
- With or without support kit (alcohol prep pad, iodophor prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandage)
- Sterile Tyvek™ packaging
- Come with Filter-Pro™ air bubble removal device
†With support kit.
Specifications
Specifications
| Needle Gauge | 22 G |
| For Use With (Application) | Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results, rapid fill minimizes patient discomfort |
| Includes | 3mL luer-lock syringe with Filter-Pro™ air bubble removal device, 22G x 1 in. preattached needle with Needle-Pro™ needle safety device, support kit (Alcohol prep pad, iodophor prep pad, gauze pad, patient label |
| Product Type | With Accessory Kit |
| Quantity | 100/Cs. |
| Packaging | Sterile Tyvek™ |
| Volume (Metric) | 3 mL |
| Sterility | Sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.