Learn More
ICU Medical Jelco™ Saf-T Holder™ Blood Tube Holders
Catalog No. 22024827
Encompass
Change view
Click to view available options
Connectors:
Female Luer
Male Luer
Product Type:
Blood Collection Tube Holder
Sterility:
Sterile
For Use With (Equipment):
Saf-T Wing, Saf-T Close
Trek Diagnostic Systems
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connectors Product Type Sterility For Use With (Equipment)
22-024-827 Female Luer Blood Collection Tube Holder Sterile Trek Diagnostic Systems
22-024-845 Female Luer Blood Collection Tube Holder Sterile Saf-T Wing, Saf-T Close
22-024-840 Male Luer Blood Collection Tube Holder Sterile Saf-T Wing, Saf-T Close
22-024-826 Male Luer Blood Collection Tube Holder Sterile Trek Diagnostic Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-024-827 Supplier ICU Medical Supplier No. 96002S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Saf-T Holder™ devices come with the back-end needle and multi-sample Luer permanently attached to the holder to minimize potential exposures and holder re-use. Available separately or with Saf-T Wing™ Blood Collection & Infusion Sets, Saf-T Holder™ devices also provide easy needless transfer after a syringe draw.
For use with Saf-T Wing™ winged infusion sets and Saf-T Closed Blood Collection System™ sets.
  • Back-end needle and multi-sample Luer permanently attached to the holder to eliminate potential exposures and holder re-use
  • Available with male or female Luer adapter
  • Sterilized with ethylene oxide
  • Latex-free
Order Info
Standard length holder fits most blood collection systems. Shorter length designed for use exclusively with Trek Diagnostic systems.
Certifications
CE marked
Specifications
For Use With (Equipment) Trek Diagnostic Systems
Product Type Blood Collection Tube Holder
Sterility Sterile
Connectors Female Luer
Latex Free Yes
For Use With (Application) Provide safe needleless transfer after a syringe draw
Certifications/Compliance CE, OSHA