Learn More
Diagnostic Biosystems SMOOTH MUSCLE MYOSIN Smooth Muscle Myosin (SMMS-1)
Supplier:  Diagnostic Biosystems MOB46705
Encompass_Preferred
Smooth muscle myosin heavy chain (SM-MHC) is a cytoplasmic structural protein which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmentally regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2, they are differentially expressed depending on the functional activity of the muscle.\nThe antibody may be useful for the study of breast tumors as the presence of an intact layer of myoepithelial cells is an important feature, which may distinguish benign breast lesions and carcinoma in situ from invasive tumors
Catalog No. 50-238-5362
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.