Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Smooth Muscle Myosin Monoclonal Antibody (SMMS-1), eBioscience™, Invitrogen™

Catalog No. 5013475
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-134-75 25 μg
50-134-76 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-134-75 Supplier Invitrogen™ Supplier No. 14640080
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This SMMS-1 monoclonal antibody reacts with both isoforms of the human smooth muscle myosin heavy chain (SM1 and SM2) structural proteins, which are components of the contractile apparatus in smooth muscle cells. Smooth muscle myosin heavy chains are expressed by myoepithelial cells and also serve as the terminal differentiation marker for smooth muscle cells. In normal breast tissue, myoepithelial cells compose the outer layer of the lumen lining. In benign and in situ breast lesions, the intact myoepithelial layer stains with SMMS-1 and the loss of this staining indicates invasive cancer. This SMMS-1 antibody has also been reported to cross-react with bovine, canine, feline, and rat smooth muscle myosin heavy chains. Applications Reported: This SMMS-1 antibody has been reported for use in western blotting, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This SMMS-1 antibody has been tested by western blot on rat heart and immunohistochemistry on formalin-fixed paraffin embedded tissue (with IHC Antigen Retrieval Solution - Low pH (cat. 00-4955)). This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be titrated for optimal perfomance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Smooth Muscle Myosin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SMMS-1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene MYH11
Gene Accession No. P35749, Q63862
Gene Alias AAT4; AV071570; contractile protein; FAA4; KIAA0866; LOW QUALITY PROTEIN: myosin-11; Myh11; myh11 {ECO:0000312; Myosin heavy chain 11; myosin heavy chain 11, smooth muscle; myosin heavy chain 21; myosin heavy chain isoform SMA; myosin heavy chain SMA; myosin heavy chain, smooth muscle isoform; myosin, heavy chain 11, smooth muscle; myosin, heavy polypeptide 11, smooth muscle; myosin-11; RGD:3136}; SM1; SM2; SMHC; SMMHC; smooth muscle myosin heavy chain
Gene Symbols MYH11
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101097693, 24582, 4629, 479836, 530050
Target Species Human, Rat, Canine, Bovine, Feline
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.