Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMPD3 Monoclonal Antibody (758612)
GREENER_CHOICE

Catalog No. MA561778
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA561778 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA561778 Supplier Invitrogen™ Supplier No. MA561778
  • $489.00
    $366.65 / Each of 1

    Save $122.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

Catalyzes the hydrolysis of sphingomyelin to form ceramide and phosphocholine. Ceramide mediates numerous cellular functions, such as apoptosis and growth arrest, and is capable of regulating these 2 cellular events independently. Also hydrolyzes sphingosylphosphocholine. Regulates the cell cycle by acting as a growth suppressor in confluent cells. Probably acts as a regulator of postnatal development and participates in bone and dentin mineralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPD3
Applications Western Blot
Classification Monoclonal
Clone 758612
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and no preservative
Gene SMPD3
Gene Accession No. Q9NY59
Gene Alias 4631433G07Rik; AI427456; AW537966; Cca1; confluent 3Y1 cell-associated 1; confluent 3Y1 cell-associated protein 1; fro; neutral sphingomyelin phosphodiesterase 3; neutral sphingomyelinase 2; Neutral sphingomyelinase II; NSMASE2; nSMase-2; SMPD3; Sphingomyelin phosphodiesterase 3; sphingomyelin phosphodiesterase 3, neutral; sphingomyelin phosphodiesterase 3, neutral membrane; sphingomyelin phosphodiesterase 3, neutral membrane (neutral sphingomyelinase II)
Gene Symbols SMPD3
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human SMPD3, Val2-Ala655.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55512
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.