Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMPD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578573
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578573 Supplier Invitrogen™ Supplier No. PA578573
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted Reactivity: Human (100%), Bovine (88%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by SMPD4 is a sphingomyelinase that catalyzes the hydrolysis of membrane sphingomyelin to form phosphorylcholine and ceramide. This gene is activated by DNA damage, cellular stress, and tumor necrosis factor, but it is downregulated by wild-type p53. The encoded protein localizes to the endoplasmic reticulum and Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPD4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SMPD4
Gene Accession No. Q6ZPR5, Q9NXE4
Gene Alias 4122402O22Rik; Kiaa1418; mKIAA1418; NET13; neutral membrane (neutral sphingomyelinase-3); neutral sphingomyelinase 3; Neutral sphingomyelinase III; neutral sphingomyelinase-3; NSMASE3; nSMase-3; RGD1310674; SKNY; SKNY protein; Smpd4; Sphingomyelin phosphodiesterase 4; sphingomyelin phosphodiesterase 4, neutral membrane; sphingomyelin phosphodiesterase 4, neutral membrane (neutral sphingomyelinase-3)
Gene Symbols SMPD4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SMPD4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303790, 55627, 77626
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.