Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMPX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3070
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3070 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3070 Supplier Invitrogen™ Supplier No. PA3070
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA3-070 detects SMPX from human, mouse, and rat samples. PA3-070 has been successfully used in Western blot, immunocytochemistry, immunohistochemistry (paraffin) procedures. By Western blot, this antibody detects a 9 kDa protein corresponding to mouse SMPX. The PA3-070 antigen is the COOH-terminal amino acids of mouse SMPX (66-84) NLSEIQNVKSELKFVPKGE coupled to KLH.

SMPX plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPX
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.02% sodium azide
Gene SMPX
Gene Accession No. Q925F0, Q9DC77, Q9UHP9
Gene Alias 1010001C09Rik; cardiac and skeletal muscle protein; chisel protein; Csl; deafness, X-linked 6, sensorineural; DFN6; DFNX4; small muscle protein X-linked; small muscle protein, X-linked; small muscular protein; Smpx; Srmx; Stretch-responsive skeletal muscle protein; Unknown (protein for MGC:128317)
Gene Symbols SMPX
Host Species Rabbit
Immunogen COOH-terminal amino acids of mouse SMPX (66-84) NLSEIQNVKSELKFVPKGE coupled to KLH.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23676, 66106, 84416
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.