Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety White SMS Disposable Anti-Static Frocks
SDP

Catalog No. 19055477
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-055-477 Medium
19-055-476 Small
19-055-478 Large
19-055-479 X-Large
19-055-480 2X-Large
19-055-482 3X-Large
19-055-483 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-055-477 Supplier Keystone Safety Supplier No. CEF3WKSMSMEDWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Frocks constructed of SMS composite fabric feature knit wrists, elastic thumb loops, and three pockets. They provide particulate and liquid barrier suitable for ISO Class 6 cleanrooms.

  • Made from SMS composite fabric
  • Disposable
  • Highly breathable and comfortable
  • ISO Class 6 cleanroom compatible
  • Low shedding and anti-static to protect sensitive products
  • Provide particulate barrier and liquid repellant
  • Ideal for food processing, laboratories, biohazards, medical, metal surface treating, pharmaceutical
  • Knit cuffs with elastic thumb loops, three pockets, six snap closure, knit collar, extra-long knee-length, hanger loop

Specifications

Size Medium
Material SMS
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Knit, Elastic Thumb Loop
Collar Type Knit
Latex Free Yes
No. of Pockets 3
Sleeve Style Long Sleeve
Packaging Type 10/Bag, 3 Bags/Lined Carton
Cleanroom Class Class 1000 (ISO 6)
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.