Learn More
Keystone Safety Blue SMS Disposable Anti-Static Frocks

Description
- Made from SMS composite fabric
- Disposable
- Highly breathable and comfortable
- ISO Class 6 cleanroom compatible
- Low shedding and anti-static to protect sensitive products
- Provide particulate barrier and liquid repellant
- Ideal for food processing, laboratories, biohazards, medical, metal surface treating, pharmaceutical
- Knit cuffs with elastic thumb loops, three pockets, six snap closure, knit collar, extra-long knee-length, hanger loop
Specifications
Specifications
| Size | X-Large |
| Material | SMS |
| Color | Blue |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Snaps |
| Cuff Style | Knit, Elastic Thumb Loop |
| Collar Type | Knit |
| Latex Free | Yes |
| No. of Pockets | 3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.