Learn More
VAI SMS Hood
GREENER_CHOICE
Catalog No. 19546553 Shop All VAI Products
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
Product Type:
Integrated Face Mask
Open Face
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Product Type
19-546-553 Large/X-Large Integrated Face Mask
19-546-552 Small/Medium Integrated Face Mask
19-546-565 Small/Medium Open Face
19-546-566 Large/X-Large Open Face
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-546-553 Supplier VAI Supplier No. 1800PHMS1825
Add to Cart
Add to Cart
This item is not returnable. View return policy
Spunbond-meltblown-spunbond (SMS) hood is available with an integrated face mask or open face design, and are individually packaged.
  • Heavyweight, non-woven and high quality microporous (MP) coated material
  • Suited for use where maximum protection to product or wearer is required
  • Light splash and chemical resistant
Specifications
Product Type Integrated Face Mask
Size Large/X-Large
Material SMS Fabric
Color White
Closure Type Pull-over
Disposable Single-use
Model 1800P-HM-S-1825
Sterility Sterile
Packaging Type Individually Packaged
Hood Type SMS