Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Blue SMS Lab Coats, Snap Front
SDP

Catalog No. 19168649
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Cuff Style:
Elastic
Knit
No. of Pockets:
3
None
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Cuff Style No. of Pockets
19-168-649 2X-Large Knit 3
19-168-644 Small Knit 3
19-812-911 Small Elastic None
19-168-646 Medium Knit 3
19-812-912 Medium Elastic None
19-168-647 Large Knit 3
19-812-913 Large Elastic None
19-168-648 X-Large Knit 3
19-812-914 X-Large Elastic None
19-812-915 2X-Large Elastic None
19-168-650 3X-Large Knit 3
19-812-916 3X-Large Elastic None
19-168-651 4X-Large Knit 3
19-812-917 4X-Large Elastic None
19-168-652 5X-Large Knit 3
19-812-918 5X-Large Elastic None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-168-649 Supplier Keystone Safety Supplier No. LC3BKSMS2XLBLUE
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Latex free SMS lab coats provide barrier protection and are comfortable to wear.

  • Latex free, 3-layer SMS fabric
  • Barrier against water based liquids and light chemical splash
  • Single collar
  • Strong, high tensile strength
  • Comfortable, lightweight, breathable
  • Serged seams
  • Meets ANSI 101-1996 and Federal Flammability ACT 16 CFR 1610.
  • Multi use for cleanroom, pharmaceutical, food, janitorial, cleaning, maintenance
  • Choose from 3 pockets or no Pockets, Elastic or Knit Wrists
  • Pack of 30

Specifications

Product Type Controlled Environments Lab Coat
Size 2X-Large
Gender Unisex
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
No. of Pockets 3
Latex Free Yes
Model LC3-BK-SMS
Certifications/Compliance Cleanroom Class 1000 and Above
For Use With (Application) Manufacturing, Electronics, Medical Device, Pharmaceutical
Autoclavable Not Autoclavable
No. per Pack 30/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.