Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ SMS Lab Coats, Elastic Wrists, No Pockets

Catalog No. 19472463 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-463 2X-Large
19-472-460 Medium
19-472-461 Large
19-472-462 X-Large
19-472-464 3X-Large
19-472-465 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-472-463 Supplier International Enviroguard™ Supplier No. 22462XL
Add to Cart
Add to Cart

Breathable SMS lab coats feature three layers of protection against particulates, light liquid splashes, or sprays.

  • Disposable SMS lab coats are lightweight, breathable, and provide a particulate barrier
  • Durable, three-layer SMS fabric protects against light liquid splashes and sprays
  • Fabric keeps wearers cool for extended periods of time
  • No pockets, elastic wrists, folded collar
  • Recommended applications include pharmaceuticals, labs, healthcare, controlled environments, and more

Specifications

Product Type Controlled Environments Lab Coat
Size 2X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
No. of Pockets None
Collar Type Folded
Seam Type Serged
For Use With (Application) Pharmaceutical, Healthcare, Controlled Environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.