Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMYD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531482 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531482 Supplier Invitrogen™ Supplier No. PA531482
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, mouse heart. Predicted reactivity: Mouse (93%), Rat (93%), Pig (95%), Chicken (83%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Acts as a transcriptional repressor. Essential for cardiomyocyte differentiation and cardiac morphogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMYD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Smyd1
Gene Accession No. P97443, Q8NB12
Gene Alias 4632404M21Rik; Bop; C78565; CD8 beta opposite; CD8beta opposite strand; CD8b-opposite; histone-lysine N-methyltransferase SMYD1; KMT3D; m-BOP; SET and MYND domain containing 1; SET and MYND domain-containing protein 1; SMYD1; Zinc finger protein BOP; zinc finger, MYND domain containing 18; Zmynd18; ZMYND22
Gene Symbols Smyd1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SMYD1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12180, 150572, 297333
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.