Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMYD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501733352 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3352 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3352 Supplier Proteintech Group Inc Supplier No. 212901AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SET domain-containing proteins, such as SMYD2, catalyze lysine methylation..

Specifications

Antigen SMYD2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SMYD2
Gene Accession No. Q8R5A0, Q9NRG4
Gene Alias HSKM B, KMT3C, Lysine N methyltransferase 3C, SMYD2, ZMYND14
Gene Symbols SMYD2
Host Species Rabbit
Immunogen SMYD2 Fusion Protein Ag15823
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226830, 56950
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.