Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMYD3 Mouse anti-Human, Clone: 1B5C10, Proteintech
SDP

Catalog No. p-7213454 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6762 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6762 Supplier Proteintech Group Inc Supplier No. 663301IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SMYD3 (histone-lysine N-methyltransferase) methylates Lys-4 of histone H3 and Lys-5 of histone H4. It induces di- and tri-methylation. SMYD3 plays an important role in transcriptional activation as a member of an RNA polymerase complex.

Specifications

Antigen SMYD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B5C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SMYD3
Gene Accession No. Q9H7B4
Gene Alias SMYD3, ZMYND1, ZNFN3A1
Gene Symbols SMYD3
Host Species Mouse
Immunogen SMYD3 Fusion Protein Ag2624
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64754
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.