Learn More
Description
The SMZ-160 is ideal for educational purposes in high schools and universities, but is also great for industrial quality control.
- Greenough Zoom Optical System
- 45° stereomicroscope head
- 6:1 zoom ratio
- Diopter Adjustment: ±5
Specifications
Specifications
| Base Style | LED Stand |
| Magnification Power | 10X |
| Objective Types | Auxiliary Objectives (0.3X, 0.5X, 0.63X, 0.75X, 1.5X, 2X) |
| Head Style | Binocular |
| Lighting Type | R2 LED |
| Eyepiece | WF 10X/20 mm |
| Working Distance | 100 mm |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.