Learn More
Motic™ SMZ 171 Stereo Microscopes
Catalog No. 01258018 Shop All Motic Group Products
Change view
Click to view available options
Head Style:
Binocular
Trinocular
Base Style:
LED Stand
Plain Stand
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style Base Style
01-258-018 Trinocular Plain Stand
01-258-016 Binocular Plain Stand
01-258-017 Binocular LED Stand
01-258-019 Trinocular LED Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 01-258-018 Supplier Motic™ Supplier No. 1100200600762
Add to Cart
Add to Cart
This item is not returnable. View return policy
The SMZ-171 model terminates Motic™’s SMZ stereo microscope line by using modern ESD-resistant materials and optimized LED illuminations for a wide range of biomedical and material science applications.
The SMZ-171 is an advanced stereo microscope for biomedical work and industrial quality control.
  • Greenough Zoom Optical System
  • 45° stereomicroscope head
  • 6.7:1 Zoom Ratio
  • Diopter Adjustment: ±5
Specifications
Base Style Plain Stand
Magnification Power 10X
Objective Types Auxiliary Objectives (0.3X, 0.5X, 0.63X, 0.75X, 1.5X, 2X)
Head Style Trinocular
Eyepiece WF 10X/23 mm
Working Distance 110 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov