Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SN 2, Tocris Bioscience™

Catalog No. 490710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-071-0 10 mg
49-075-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-071-0 Supplier R&D Systems Supplier No. 4907/10
This item is not returnable. View return policy

Selective TRPML3 channel activator

Selective TRPML3 channel activator (EC50 = 1.13 μM).

Chemical Identifiers

CAS 823218-99-1
Molecular Formula C17H21NO
Molecular Weight (g/mol) 255.361
InChI Key WKLZNTYMDOPBSE-UHFFFAOYSA-N
Synonym sn 2, sn-2, 5-mesityl-3-oxa-4-azatricyclo 5.2.1.0∼2,6∼ dec-4-ene, 5-mesityl-3-oxa-4-azatricyclo 5.2.1.02,6 dec-4-ene, d0g1ln, 5-mesityl-3-oxa-4-azatricyclo 5.2.1.0 2,6 dec-4-ene, 3a,4,5,6,7,7a-hexahydro-3-2,4,6-trimethylphenyl-4,7-methano-1,2-benzisoxazole
PubChem CID 11834987
SMILES CC1=CC(=C(C(=C1)C)C2=NOC3C2C4CCC3C4)C

Specifications

Color White
Quantity 10 mg
Formula Weight 255.35
Percent Purity >98%
Physical Form Solid
Chemical Name or Material SN 2
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.