Learn More
SNAI2 Mouse anti-Human, Clone: OTI2C3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504933

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Slug, a member of the Snail family of C2H2-type zinc finger transcription factors, was initially identified to be involved in epithelial-mesenchymal transitions as well as the formation of the neural tube during vertebrate embryogenesis. Like Snail, Slug transcription can be induced by growth factors such as FGF, BMP, and TGF-beta. Once expressed, Slug will bind E-box regions of promoters and repress transcription of genes such as E-cadherin and Claudin-1. More recently, its expression in breast, esophogeal, and colorectal carcinomas has been correlated with poor prognosis for survival. Furthermore, Slug can protect hemapoietic progenitor cells from radiation-induced apoptosis by repressing the p53-mediated transcription of Puma, a BH3-only antagonist of the anti-apoptotic members of the Bcl-2 family.Specifications
SNAI2 | |
Monoclonal | |
Unconjugated | |
SNAI2 | |
SLUG, SLUGH, SLUGH1, SNAIL2, WS2D | |
Mouse | |
Affinity Chromatography | |
RUO | |
6591 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI2C3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O43623 | |
SNAI2 | |
Full length human recombit protein of human SNAI2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.