Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAI3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595254
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595254 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595254 Supplier Invitrogen™ Supplier No. PA595254
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: MCF-7 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

SNAI3 is a member of the SNAIL gene family, named for the Drosophila snail gene, which plays roles in mesodermal formation during embryogenesis (Katoh and Katoh, 2003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAI3
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene SNAI3
Gene Accession No. Q3KNW1
Gene Alias AI643946; Protein snail homolog 3; Smuc; SNAI3; snail family transcriptional repressor 3; snail family zinc finger 3; snail homolog 3; SNAIL3; snail-related gene from muscle cells; snail-related protein 2; Zfp293; zinc finger protein 293; Zinc finger protein SNAI3; ZNF293
Gene Symbols SNAI3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human SNAI3 (155-178aa FHCHKPYHTLAGLARHRQLHCHLQ).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 333929
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.