Learn More
Cayman Chemical SNAP, 67776-06-1, 10 mg
SDP
Supplier:  Cayman Chemical 8225010
Encompass_Preferred
Molecular Formula C7H12N2O4S, Purity ≥98%, Formula Weight 220.2, Synonyms: S-Nitroso-N-Acetyl-D,L-Penicillamine; A commonly used NO donor and a potent vasodilator.
Catalog No. NC9701277
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.