Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAP alpha Monoclonal Antibody (15D4)
GREENER_CHOICE

Catalog No. 01674320
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-320 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-320 Supplier Invitrogen™ Supplier No. MA112453
  • $599.00
    $497.30 / Each of 1

    Save $101.70

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SNAP alpha Monoclonal Antibody for Western Blot, IP, ELISA

SNAPs (soluble NSF attachment proteins), acting in concert with SNAREs (SNAP receptors) and the N-ethylmaleimide-sensitive fusion protein (NSF) are required for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-to-endosome fusion and transcytotic vesicles-to-plasma membrane transport. Vesicle-to-target membrane docking (initial contact) occurs when the vesicle SNARE binds to its cognate target membrane SNARE. a-SNAP (or b-SNAP in brain) then binds to this docking complex and mediates the binding of NSF and thus the formation of a 20S fusion particle. It is thought that, once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP alpha
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 15D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and no preservative; pH 7.5
Gene NAPA
Gene Accession No. P54920, P54921, P81125
Gene Alias 1500039N14Rik; alpha snap; alpha soluble NSF attachment protein; alpha-SNAP; Alpha-soluble NSF attachment protein; a-SNAP; AW209189; hydrocephaly with hop gait; hyh; Napa; N-ethylmaleimide sensitive fusion protein attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein, alpha; NSF attachment protein alpha; RA81; Snap; SNAPA; SNAP-alpha; SNARE
Gene Symbols NAPA
Host Species Mouse
Immunogen Recombinant human alpha-SNAP.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140673, 518407, 8775
Target Species Human, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.