Learn More
ProteinSimple SNAP BURST CAP 11MM
Supplier:  ProteinSimple 102031
Encompass
Snap burst cap - 11mm with septum/Si, 100/pkg. Suitable for use with all iCE280 and iCE3 configurations.
Catalog No. NC9518932