Learn More
Thermo Scientific™ Snap-Cap Centrifuge Tubes, Plain
Catalog No. 21403175 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Natural
Sterility:
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility
21-403-175 Natural Sterile
21-403-161 Natural Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-175 Supplier Thermo Scientific™ Supplier No. 3443
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Thermo Scientific™ Plain Snap-Cap Centrifuge Tubes include a flat cap, and have no frosting or graduations.
  • With flat cap
  • No frosting or graduations
  • Certified RNase/DNase, RNA/DNA, ATP and pyrogen free
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Flat
Certifications/Compliance Certified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description Plain, Snap-Cap Centrifuge Tube
Capacity (Metric) 1.5 mL
Color Natural
Material Polypropylene
Packaging 10 bags of 50
Sterility Sterile
Graduated No
Max. RCF 25,000 x G