Learn More
Description
- Features raised tab for easy removal of cap
- Provides leak-resistant seal
Specifications
Specifications
| Product Type | Snap Cap |
| Autoclavable | Not Autoclavable |
| Color | Yellow |
| No. per Case | 1500 |
| Sterility | Non-sterile |
| For Use With (Equipment) | Flared Top Urine Tubes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.