Learn More
Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes
GREENER_CHOICE
Catalog No. 21402903 Shop All Thermo Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Capacity (Metric):
0.6 mL
1.5 mL
2 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Capacity (Metric)
21-402-903 Sterile 1.5 mL
21-402-905 Sterile 2 mL
21-402-904 Non-sterile 2 mL
21-403-189 Non-sterile 0.6 mL
21-403-190 Sterile 0.6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 21-402-903 Supplier Thermo Scientific™ Supplier No. 3451
Add to Cart
Edge
Add to Cart
Improve sample recovery by significantly reducing surface binding with these tubes that do not have additives or coatings.
Improve sample recovery by significantly reducing surface binding with Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes. Viscous liquids and costly reagents retained in a microcentrifuge tube effect assay accuracy and precision. With no additives or coatings, these tubes — available in 0.6mL, 1.5mL and 2.0mL sizes — offer maximum sample recovery by significantly reducing surface binding.
  • The duckbill cap reduces the pressure required to open the tube with a wider contact surface, yet ensures the cap will remain closed during centrifugation
  • Low retention properties are not achieved by a coating or plastic-embedding processes, ensuring that samples will not be contaminated from these additives
  • Tubes will remain closed during centrifugation yet open easily with one hand as needed
  • Tubes are available in amber to protect your light-sensitive samples
  • Graduation marks on the microcentrifuge tube allow for effective sample reference
Highest Quality
  • Manufactured from virgin polypropylene, these microcentrifuge tubes ensure sample integrity during boiling, freezing and centrifugation
  • Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
  • Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
Recommended for:
Daily laboratory usage including centrifugation and short-term sample storage
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Flat
Certifications/Compliance Certified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Capacity (Metric) 1.5 mL
Color Natural
Material Polypropylene
Packaging 50 tubes/ bag, 10 bags/ unit, 10 units/ case
Sterility Sterile
Graduated Yes
Max. RCF 26,000 x g