Learn More
Description
- Designed specifically for use on flared-top urine centrifuge tubes
- Raised tab allows for easy removal of the cap
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Polyethylene |
| Color | Blue |
| For Use With (Equipment) | Flared Top Urine Tubes |
| Disposable | Single-use |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.