Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG™ Snap-Over LDPE Bucket Insert
SDP

Catalog No. 18366513 Shop All New Pig Corporation Products
Change view
Click to view available options
Capacity (English):
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18-366-513 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-513 Supplier PIG™ Supplier No. DRM255
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FDA Compliant pail liners that snap over pail rims to reduce chance of leaks.

  • Thin, contoured lip snaps over pail rim to reduce chance of leaks, while still allowing a tight seal with lid
  • Made of FDA compliant polyethylene, safe for use with food and cosmetic products
  • Helps eliminate container residues
  • Ideal for storing paints, inks, coatings and other hard-to-clean materials
  • Seamless, LDPE construction is chemical resistant and eliminates the chance of seam leaks
  • For use with 5 gal. poly buckets
  • Thickness: 15 mil
  • Quantity: 50 per box

Specifications

Product Type Drum Liner
Capacity (English) 5 gal.
Capacity (Metric) 18.92 L
Color Clear
For Use With (Equipment) 5 gal. Poly Bucket
Material LDPE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.