Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAP25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674382 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-382 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-382 Supplier Invitrogen™ Supplier No. PA19102
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

SNAP25 is a t-SNARE involved in the molecular regulation of neurotransmitter release. It may play a role in the synaptic function of specific neuronal systems. SNAP25 associates with proteins involved in vesicle docking and membrane fusion. It also regulates plasma membrane recycling through its interaction with CENPF. Mutations in the gene can results in myasthenic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP25
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Snap25
Gene Accession No. P60879, P60880, P60881
Gene Alias ALMS1; bA416N4.2; Bdr; blind drunk; CG17676; CG17884; CG40452; CMS18; dJ1068F16.2; Dmel CG40452; Dmel_CG40452; etID17571.20; etID69167.20; fj42b02; fj45g04; fj48c02; FLJ23079; GENA70; HGNC:11132; resistance to inhibitors of cholinesterase 4 homolog; RIC4; RIC-4; SEC9; si:dkeyp-8f4.6; Snap; SNAP 25; snap25; SNAP-25; snap25.1; SNAP-25.1; snap25a; SNAP-25A; SNAP-25B; snp25; sp; SUP; super protein; synaptosomal associated protein-25; synaptosomal-associated 25 kDa protein; Synaptosomal-associated protein 25; synaptosomal-associated protein 25-A; synaptosomal-associated protein, 25 kDa; synaptosomal-associated protein, 25a; synaptosomal-associated protein, 25kDa; synaptosome associated protein 25; synaptosome associated protein 25a; synaptosome associated protein 25kDa; synaptosome-associated protein 25.1; synaptosome-associated protein 25a; wu:fj42b02; wu:fj45g04; wu:fj48c02
Gene Symbols Snap25
Host Species Goat
Immunogen Synthetic peptide corresponding to the C terminus amino acids DEANQRATKMLGSG (aa 193-206).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20614, 25012, 6616
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.